Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,4-Triazin-3-amine, 97%, Maybridge

Click to view available options
Quantity:
1g
5g
1120-99-6
C3H4N4
96.093
MJIWQHRXSLOUJN-UHFFFAOYSA-N
3-amino-1,2,4-triazine,as-triazine, 3-amino,3-amino-as-triazine,1,2,4-triazin-3-ylamine,1,2,4-triazine-3-ylamine,1,2,4 triazin-3-ylamine,zlchem 1277,1,2,4-triazine-3-amine,3-amino-1,2,4,-triazine
70715
1,2,4-triazin-3-amine
C1=CN=NC(=N1)N
1120-99-6
C3H4N4
96.093
MJIWQHRXSLOUJN-UHFFFAOYSA-N
3-amino-1,2,4-triazine,as-triazine, 3-amino,3-amino-as-triazine,1,2,4-triazin-3-ylamine,1,2,4-triazine-3-ylamine,1,2,4 triazin-3-ylamine,zlchem 1277,1,2,4-triazine-3-amine,3-amino-1,2,4,-triazine
70715
1,2,4-triazin-3-amine
C1=CN=NC(=N1)N
Quantity 1g
Assay Percent Range 97%
Formula Weight 96.09
Chemical Name or Material 1, 2, 4-Triazin-3-amine