Learn More
1,2,4-Triazole, 99%, Thermo Scientific Chemicals
Description
1,2,4-Triazole is widely used as an intermediate in the manufacture of pharmaceuticals. It acts as an anti-scaling agent and antifungal compound. It is widely used in antifungals such as fluconazole anditraconazole.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,2,4-Triazole is widely used as an intermediate in the manufacture of pharmaceuticals. It acts as an anti-scaling agent and antifungal compound. It is widely used in antifungals such as fluconazole anditraconazole.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents and bases.
Specifications
Melting Point | 118°C to 122°C |
Boiling Point | 260°C |
Flash Point | 158°C (316°F) |
Quantity | 25 g |
Beilstein | 104767 |
Merck Index | 14,9605 |
Solubility Information | Soluble in water. |
Formula Weight | 69.07 |
Percent Purity | 99% |
Chemical Name or Material | 1,2,4-Triazole |
Safety and Handling
Hazard Category | H302-H319-H361d |
Hazard Statement | GHS H Statement H361-H302-H319 Suspected of damaging fertility or the unborn child. Harmful if swallowed. Causes serious eye irritation. |
Precautionary Statement | P201-P202-P264b-P270-P280i-P281-P301+P312-P305+P351+P338-P308+P313-P330-P501c |
EINECSNumber | 206-022-9 |
RTECSNumber | XZ3806000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.