Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,4-Triazole, sodium derivative, 90%, Tech., ACROS Organics™

Click to view available options
Quantity:
10g
50g
250g
Packaging:
Glass bottle
41253-21-8
C2H2N3Na
91.05
MFCD00044437
NVMNEWNGLGACBB-UHFFFAOYSA-N
sodium 1,2,4-triazol-1-ide,1,2,4-triazolylsodium,1,2,4-triazole sodium derivative,1,2,4-triazole sodium salt,sodium 1,2,4-triazolide,1h-1,2,4-triazole, sodium salt,sodium 1h-1,2,4-triazolate,1,2,4-triazole sodium,sodium 1h-1,2,4-triazolide,1,2,4-triazole, sodium salt
4649906
sodium;2,4-diaza-1-azanidacyclopenta-2,4-diene
C1=NC=N[N-]1.[Na+]
41253-21-8
C2H2N3Na
91.05
MFCD00044437
NVMNEWNGLGACBB-UHFFFAOYSA-N
sodium 1,2,4-triazol-1-ide,1,2,4-triazolylsodium,1,2,4-triazole sodium derivative,1,2,4-triazole sodium salt,sodium 1,2,4-triazolide,1h-1,2,4-triazole, sodium salt,sodium 1h-1,2,4-triazolate,1,2,4-triazole sodium,sodium 1h-1,2,4-triazolide,1,2,4-triazole, sodium salt
4649906
sodium;2,4-diaza-1-azanidacyclopenta-2,4-diene
C1=NC=N[N-]1.[Na+]
Melting Point 295°C
Infrared Spectrum Authentic
Identification (sodium): Passes Test
Packaging Glass bottle
Quantity 50g
Color White to Brown
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 90%
Beilstein 26, II, 7
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in most organic solvents
Formula Weight 91.05
Physical Form Crystalline Powder
Percent Purity 90%
Grade Technical
Chemical Name or Material 1,2,4-Triazole, sodium derivative, tech., 90%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes serious eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

GHS Signal Word: Danger

EINECSNumber : 255-280-9

TSCA : TSCA