Learn More
1,2,4-Trichlorobenzene, Spectrophotometric Grade, 99% min, Thermo Scientific Chemicals
Description
1,2,4-Trichlorobenzene is used as a dielectric and heat transfer fluid in transformers. It acts as an intermediate, degreaser, wood preservative and solvent for dye. It is a high-temperature solvent used in gel permeation chromatography, especially for polyethylene and polypropylene. Further, it is used as a lubricant and as a synthetic transformer oil.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 17°C |
Color | Colorless |
Density | 1.454 |
Boiling Point | 213°C |
Quantity | 250 mL |
Flash Point | 110°C (230°F) |
Ignition Point | 571°C (1060°F) |
Odor | Aromatic |
Physical Form | Liquid |
Refractive Index | 1.572 |
Show More |
Safety and Handling
Hazard Category | H302-H315-H319 |
Precautionary Statement | P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c |
DOTInformation | DOT Class: 6.1, Packaging Group: III |
EINECSNumber | 204-428-0 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.