Alfa Aesar™ 1,2,4-Trifluorobenzene, 98+%

Manufacturer: Alfa Aesar™ A1142422

Quantity 100g

 View more versions of this product

Catalog No. AAA1142422

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 367-23-7
    Molecular Formula C6H3F3
    Formula Weight 132.09
    MDL Number MFCD00000305
    UN Number UN2387
    Beilstein 2042865
    Chemical Name or Material 1,2,4-Trifluorobenzene

    Specifications

    Formula Weight 132.09
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 89° to 91°C
    Molecular Formula C6H3F3
    MDL Number MFCD00000305
    Beilstein 2042865
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLUOROBENZENE
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H315-H319-H335
    Highly flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    CAS 367-23-7
    UN Number UN2387
    Flash Point 4°C (39°F)
    Quantity 100g
    Chemical Name or Material 1,2,4-Trifluorobenzene
    Assay ≥98%

    GHS H Statement
    H225-H315-H319-H335
    Highly flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit