Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,4-Trimethylbenzene, 98%, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
10L
25L
Packaging:
Glass Bottle
Plastic drum
95-63-6
C9H12
120.19
MFCD00008527
GWHJZXXIDMPWGX-UHFFFAOYSA-N
pseudocumene,pseudocumol,psi-cumene,as-trimethylbenzene,1,3,4-trimethylbenzene,benzene, 1,2,4-trimethyl,uns-trimethylbenzene,1,2,5-trimethylbenzene,asymmetrical trimethylbenzene,.psi.-cumene
7247
CHEBI:34039
1,2,4-trimethylbenzene
CC1=CC(=C(C=C1)C)C
95-63-6
C9H12
120.19
MFCD00008527
GWHJZXXIDMPWGX-UHFFFAOYSA-N
pseudocumene,pseudocumol,psi-cumene,as-trimethylbenzene,1,3,4-trimethylbenzene,benzene, 1,2,4-trimethyl,uns-trimethylbenzene,1,2,5-trimethylbenzene,asymmetrical trimethylbenzene,.psi.-cumene
7247
CHEBI:34039
1,2,4-trimethylbenzene
CC1=CC(=C(C=C1)C)C
Specific Gravity 0.88
Melting Point -44.0°C
Density 0.8800g/mL
Infrared Spectrum Authentic
Flash Point 48°C
Refractive Index 1.5030 to 1.5050
Packaging Plastic drum
Boiling Point 168.0°C
Quantity 25L
Color Colorless
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C6H3(CH3)3
Beilstein 05,4
Merck Index 15,8023
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, benzene and ether
Formula Weight 120.19
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 1,2,4-Trimethylbenzene
Show More Show Less

GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
Toxic to aquatic life with long lasting effects.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning