Alfa Aesar™ 1-(2,6-Difluorophenyl)ethanol, 97%

Manufacturer: Alfa Aesar™ H3277103

Quantity 1g

 View more versions of this product

Catalog No. AAH3277103

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 87327-65-9
    Molecular Formula C8H8F2O
    Formula Weight 158.15
    MDL Number MFCD00274321
    Chemical Name or Material 1-(2,6-Difluorophenyl)ethanol

    Specifications

    Formula Weight 158.15
    Molecular Formula C8H8F2O
    MDL Number MFCD00274321
    CAS 87327-65-9
    Quantity 1g
    Chemical Name or Material 1-(2,6-Difluorophenyl)ethanol
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit