Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,6-Hexanetriol, 97+%, extra pure, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
106-69-4
C6H14O3
134.175
MFCD00002976
ZWVMLYRJXORSEP-UHFFFAOYSA-N
1,2,6-hexanetriol,1,2,6-trihydroxyhexane,hexanetriol-1,2,6,1,6-hexanetriol,hexanetriol-1,6,hexane-1,6-triol,1,6-trihydroxyhexane,acmc-1bsub,dsstox_cid_21224
7823
hexane-1,2,6-triol
C(CCO)CC(CO)O
106-69-4
C6H14O3
134.175
MFCD00002976
ZWVMLYRJXORSEP-UHFFFAOYSA-N
1,2,6-hexanetriol,1,2,6-trihydroxyhexane,hexanetriol-1,2,6,1,6-hexanetriol,hexanetriol-1,6,hexane-1,6-triol,1,6-trihydroxyhexane,acmc-1bsub,dsstox_cid_21224
7823
hexane-1,2,6-triol
C(CCO)CC(CO)O
Color Yellow
Appearance (10% in ethanol) clear, slightly yellow
Packaging Glass bottle
Refractive Index 1.4750 to 1.4770 (20°C, 589nm)
Water 0.2% max. (K.F.)
pH 4.0 to 7.0 (30% soln. at 20°C)
Quantity 5g
Infrared Spectrum Authentic
Density 1.1090g/mL
CAS Min % 97.0
CAS Max % 100.0
Assay Percent Range 97+%
Linear Formula HO(CH2)4CH(OH)CH2OH
Formula Weight 134.18
Physical Form Viscous Liquid
Percent Purity ≥97.0%
Grade Extra Pure
Chemical Name or Material 1, 2, 6-Hexanetriol, 97%
Show More Show Less

HYGROSCOPIC

EINECSNumber : 203-424-6

RTECSNumber : MO4650000

TSCA : TSCA

Recommended Storage : the product may solidify at ca. 32°C