Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-(2-Aminoethyl)-2-methyl-5-nitroimidazole dihydrochloride monohydrate, 98%, ACROS Organics™
Click to view available options
Quantity:
5g
Specifications
Melting Point | 195°C |
Packaging | Glass bottle |
Quantity | 5g |
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Assay Percent Range | 98% |
Solubility Information | Solubility in water: 600g/L. Other solubilities: methanol:26g/L ethanol:1.8g/L 2-propanol:0.48g/l,dichloromethane: 0.11g/L,acetone: 0.11g/L,n,n-dimethylformamide: 7g/L |
Formula Weight | 261.11 |
Percent Purity | 98% |
Chemical Name or Material | 1-(2-Aminoethyl)-2-methyl-5-nitroimidazole dihydrochloride monohydrate, 98% |
Safety and Handling
GHS H Statement
Suspected of causing cancer.
Harmful if swallowed.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Obtain special instructions before use.
GHS Signal Word: Warning
EINECSNumber : 256-385-2
TSCA : TSCA