Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Aminophenyl)pyrrole, 98+%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
6025-60-1
C10H10N2
158.204
MFCD00005344
GDMZHPUPLWQIBD-UHFFFAOYSA-N
1-2-aminophenyl pyrrole,2-1h-pyrrol-1-yl aniline,n-2-aminophenyl pyrrole,2-1h-pyrrol-1-yl phenyl amine,1-2-aminophenyl-1h-pyrrole,2-1-pyrrolyl aniline,benzenamine, 2-1h-pyrrol-1-yl,2-pyrrolylphenylamine,2-pyrrol-1-yl aniline,acmc-20apd3
80123
2-pyrrol-1-ylaniline
C1=CC=C(C(=C1)N)N2C=CC=C2
6025-60-1
C10H10N2
158.204
MFCD00005344
GDMZHPUPLWQIBD-UHFFFAOYSA-N
1-2-aminophenyl pyrrole,2-1h-pyrrol-1-yl aniline,n-2-aminophenyl pyrrole,2-1h-pyrrol-1-yl phenyl amine,1-2-aminophenyl-1h-pyrrole,2-1-pyrrolyl aniline,benzenamine, 2-1h-pyrrol-1-yl,2-pyrrolylphenylamine,2-pyrrol-1-yl aniline,acmc-20apd3
80123
2-pyrrol-1-ylaniline
C1=CC=C(C(=C1)N)N2C=CC=C2
Melting Point 94°C to 98°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5g
Color Beige to Brown
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Formula Weight 158.2
Physical Form Crystalline Powder and Needles
Percent Purity ≥98.0%
Chemical Name or Material 1-(2-Aminophenyl)pyrrole, 98+%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 227-884-2

TSCA : TSCA