Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,2-Benzenedimethanol, 97%

Click to view available options
Quantity:
1g
5g
25g
612-14-6
C8H10O2
138.166
MFCD00004626
XMUZQOKACOLCSS-UHFFFAOYSA-N
1,2-benzenedimethanol,1,2-phenylenedimethanol,phthalyl alcohol,1,2-dihydroxymethylbenzene,o-xylene-alpha,alpha'-diol,2-hydroxymethyl phenyl methanol,benzenedimethanol,o-xylylene glycol,1,2-di hydroxymethyl benzene,2-hydroxymethylbenzenemethanol
69153
[2-(hydroxymethyl)phenyl]methanol
C1=CC=C(C(=C1)CO)CO
612-14-6
C8H10O2
138.166
MFCD00004626
XMUZQOKACOLCSS-UHFFFAOYSA-N
1,2-benzenedimethanol,1,2-phenylenedimethanol,phthalyl alcohol,1,2-dihydroxymethylbenzene,o-xylene-alpha,alpha'-diol,2-hydroxymethyl phenyl methanol,benzenedimethanol,o-xylylene glycol,1,2-di hydroxymethyl benzene,2-hydroxymethylbenzenemethanol
69153
[2-(hydroxymethyl)phenyl]methanol
C1=CC=C(C(=C1)CO)CO
Applications
1,2-Benzenedimethanol is a raw material used in chemical synthesis.

Solubility
Soluble in water, ether, ethanol, benzene.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
Quantity 5g
Melting Point 59°C to 64°C
Beilstein 508775
Formula Weight 138.17
Percent Purity 97%
Chemical Name or Material 1,2-Benzenedimethanol