Alfa Aesar™ 1,2-Bis(2-chloroethoxy)ethane, 97%

Manufacturer: Alfa Aesar™ B2288436

Quantity 500g

 View more versions of this product

Catalog No. AAB2288436

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 112-26-5
    Molecular Formula ClCH2CH2OCH2CH2OCH2CH2Cl
    Formula Weight 187.07
    MDL Number MFCD00000976
    UN Number UN2810
    Beilstein 506177
    Synonym Triethylene glycol dichloride
    Chemical Name or Material 1,2-Bis(2-chloroethoxy)ethane

    Specifications

    Formula Weight 187.07
    TSCA TSCA
    Molecular Formula ClCH2CH2OCH2CH2OCH2CH2Cl
    MDL Number MFCD00000976
    UN Number UN2810
    Melting Point −32°C
    Boiling Point 235°C
    Synonym Triethylene glycol dichloride
    Quantity 500g
    Flash Point >110°C (230°F)
    Health Hazard 2 GHS H Statement
    H302-H312-H315-H335
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P304+P340-P362-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Take off contaminated clothing and wash before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number KH4900000
    CAS 112-26-5
    Beilstein 506177
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
    Chemical Name or Material 1,2-Bis(2-chloroethoxy)ethane
    Assay 97%

    Description

    Applications
    1,2-Bis(2-chloroethoxy)ethane is used as pharmaceutical intermediate.

    Solubility
    Insoluble in water.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    GHS H Statement
    H302-H312-H315-H335
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P304+P340-P362-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Take off contaminated clothing and wash before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit