Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,2-Bis(2-nitrophenoxy)ethane, 98%

Click to view available options
Quantity:
10g
50g
250g
51661-19-9
C14H12N2O6
304.258
MFCD00024226
DEHTVRKGDUCXRF-UHFFFAOYSA-N
1,2-bis o-nitrophenoxy ethane,1,2-bis 2-nitrophenoxy ethane,1-nitro-2-2-2-nitrophenoxy ethoxy benzene,ethylene glycol bis 2-nitrophenyl ether,1,1'-ethane-1,2-diylbis oxy bis 2-nitrobenzene,bis 2-nitrophenoxy ethane,acmc-20a59z,1,2-di o-nitrophenoxy ethane,2,2'-ethylenedioxy bis 1-nitrobenzene,2-nitro-1-2-2-nitrophenoxy ethoxy benzene
394580
1-nitro-2-[2-(2-nitrophenoxy)ethoxy]benzene
C1=CC=C(C(=C1)[N+](=O)[O-])OCCOC2=CC=CC=C2[N+](=O)[O-]
51661-19-9
C14H12N2O6
304.258
MFCD00024226
DEHTVRKGDUCXRF-UHFFFAOYSA-N
1,2-bis o-nitrophenoxy ethane,1,2-bis 2-nitrophenoxy ethane,1-nitro-2-2-2-nitrophenoxy ethoxy benzene,ethylene glycol bis 2-nitrophenyl ether,1,1'-ethane-1,2-diylbis oxy bis 2-nitrobenzene,bis 2-nitrophenoxy ethane,acmc-20a59z,1,2-di o-nitrophenoxy ethane,2,2'-ethylenedioxy bis 1-nitrobenzene,2-nitro-1-2-2-nitrophenoxy ethoxy benzene
394580
1-nitro-2-[2-(2-nitrophenoxy)ethoxy]benzene
C1=CC=C(C(=C1)[N+](=O)[O-])OCCOC2=CC=CC=C2[N+](=O)[O-]
Melting Point 167°C to 170°C
Quantity 10g
Formula Weight 304.26
Percent Purity 98%
Chemical Name or Material 1,2-Bis(2-nitrophenoxy)ethane

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning