Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis[2-(trifluoromethyl)phenyl]ethane, 97+%, Alfa Aesar™

Click to view available options
Quantity:
1g
5g
287172-66-1
C16H12F6
318.262
MFCD01631639
OBMDBKNYJGWWJX-UHFFFAOYSA-N
1,2-bis 2'-trifluoromethyl phenyl ethane,1-trifluoromethyl-2-2-2-trifluoromethyl phenyl ethyl benzene,1,2-bis-2-trifluoromethylphenyl ethane,1,2-bis-2-trifluoromethyl phenyl ethane,1,2-bis 2 trifluoromethyl phenyl ethane
2773232
1-(trifluoromethyl)-2-[2-[2-(trifluoromethyl)phenyl]ethyl]benzene
C1=CC=C(C(=C1)CCC2=CC=CC=C2C(F)(F)F)C(F)(F)F
287172-66-1
C16H12F6
318.262
MFCD01631639
OBMDBKNYJGWWJX-UHFFFAOYSA-N
1,2-bis 2'-trifluoromethyl phenyl ethane,1-trifluoromethyl-2-2-2-trifluoromethyl phenyl ethyl benzene,1,2-bis-2-trifluoromethylphenyl ethane,1,2-bis-2-trifluoromethyl phenyl ethane,1,2-bis 2 trifluoromethyl phenyl ethane
2773232
1-(trifluoromethyl)-2-[2-[2-(trifluoromethyl)phenyl]ethyl]benzene
C1=CC=C(C(=C1)CCC2=CC=CC=C2C(F)(F)F)C(F)(F)F
Quantity 1g
Melting Point 74°C to 76°C
Formula Weight 318.26
Percent Purity ≥97%
Chemical Name or Material 1,2-Bis[2-(trifluoromethyl)phenyl]ethane

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning