Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-1,2-Bis[(2R,5R)-2,5-diethyl-1-phospholanyl]ethane, 97+%, Thermo Scientific™

Click to view available options
Quantity:
250mg
1g
136705-62-9
C18H36P2
314.434
MFCD01073768
QOLRLVPABLMMKI-BRSBDYLESA-N
et-bpe, r,r, r,r-et-bpe, +-1,2-bis 2r,5r-2,5-diethylphospholano ethane, r,r-et-bpe mi, bpe ligands r,r-et-bpe mi, phospholane, 1,1'-1,2-ethanediyl bis 2,5-diethyl-, 2r,2'r,5r,5'r, 1,2-bis 2r,5r-2,5-diethylphospholan-1-yl ethane, 1,1'-ethylenebis 2alpha,5beta-diethyl-1-phosphacyclopentane
11433485
(2R,5R)-1-[2-[(2R,5R)-2,5-diethylphospholan-1-yl]ethyl]-2,5-diethylphospholane
CCC1CCC(P1CCP2C(CCC2CC)CC)CC
136705-62-9
C18H36P2
314.434
MFCD01073768
QOLRLVPABLMMKI-BRSBDYLESA-N
et-bpe, r,r, r,r-et-bpe, +-1,2-bis 2r,5r-2,5-diethylphospholano ethane, r,r-et-bpe mi, bpe ligands r,r-et-bpe mi, phospholane, 1,1'-1,2-ethanediyl bis 2,5-diethyl-, 2r,2'r,5r,5'r, 1,2-bis 2r,5r-2,5-diethylphospholan-1-yl ethane, 1,1'-ethylenebis 2alpha,5beta-diethyl-1-phosphacyclopentane
11433485
(2R,5R)-1-[2-[(2R,5R)-2,5-diethylphospholan-1-yl]ethyl]-2,5-diethylphospholane
CCC1CCC(P1CCP2C(CCC2CC)CC)CC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Density 0.939
Boiling Point 104°C to 106°C (0.05mmHg)
Flash Point 110°C (230°F)
UN Number UN2845
Refractive Index 1.525
Quantity 250mg
Sensitivity Air Sensitive
Formula Weight 314.43
Optical Rotation +320° (c=1 in Hexane)
Percent Purity ≥97%
Chemical Name or Material (+)-1,2-Bis[(2R,5R)-2,5-diethyl-1-phospholanyl]ethane
Show More Show Less

GHS H Statement
H250-H252-H315-H319-H335
Catches fire spontaneously if exposed to air.
Self-heating in large quantities; may catch fire.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P222-P235+P410-P261-P264b-P271-P280-P302+P334-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H250-H252-H315-H319-H335

DOTInformation : Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

RUO – Research Use Only