Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-1,2-Bis[(2R,5R)-2,5-dimethylphospholano]ethane, 95%, ACROS Organics™

Click to view available options
Quantity:
100mg
Packaging:
Glass Bottle in Plastic Container
129648-07-3
C14H28P2
258.326
MFCD01073770
IRCDUOCGSIGEAI-AAVRWANBSA-N
r,r-me-bpe,unii-1ls1afh1f1,1ls1afh1f1,+-1,2-bis 2r,5r-2,5-dimethylphospholano ethane,s,s-me-en-duphos,me-bpe, r,r,r,r-me-bpe mi,r,r-me-bpe, +,2r,5r-1-2-2r,5r-2,5-dimethylphospholan-1-yl ethyl-2,5-dimethylphospholane
2734555
(2R,5R)-1-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]ethyl]-2,5-dimethylphospholane
CC1CCC(P1CCP2C(CCC2C)C)C
129648-07-3
C14H28P2
258.326
MFCD01073770
IRCDUOCGSIGEAI-AAVRWANBSA-N
r,r-me-bpe,unii-1ls1afh1f1,1ls1afh1f1,+-1,2-bis 2r,5r-2,5-dimethylphospholano ethane,s,s-me-en-duphos,me-bpe, r,r,r,r-me-bpe mi,r,r-me-bpe, +,2r,5r-1-2-2r,5r-2,5-dimethylphospholan-1-yl ethyl-2,5-dimethylphospholane
2734555
(2R,5R)-1-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]ethyl]-2,5-dimethylphospholane
CC1CCC(P1CCP2C(CCC2C)C)C
Color Colorless
Quantity 100mg
Refractive Index 1.5330 to 1.5350 (20°C, 589nm)
Packaging Glass Bottle in Plastic Container
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Merck Index 15, 1363
Formula Weight 258.33
Physical Form Oily Liquid
Percent Purity ≥94%
Chemical Name or Material (+)-1,2-Bis[(2R,5R)-2,5-dimethylphospholano]ethane
Show More Show Less