Alfa Aesar™ (+)-1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) trifluoromethanesulfonate

Catalytic asymmetric cycloisomerization and enantio selections hydrogenation reactions

Manufacturer: Alfa Aesar™ 44546TF

Quantity 0.05g

 View more versions of this product

Catalog No. AA44546TF

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 142184-30-3
    Molecular Formula Rh(C8H12)(C22H36P2)CF3SO3
    Formula Weight 722.63
    MDL Number MFCD00269861
    Chemical Name or Material (+)-1,2-Bis ( (2S,5S)-2,5-diethylphospholano)benzene (cyclooctadiene)rhodium (I) trifluoromethanesulfonate

    Specifications

    Formula Weight 722.63
    Physical Form Crystalline
    Molecular Formula Rh(C8H12)(C22H36P2)CF3SO3
    MDL Number MFCD00269861
    Quantity 0.05g
    Sensitivity Air & Moisture Sensitive
    CAS 142184-30-3
    Storage Note 1
    • Packaged under Argon
    Chemical Name or Material (+)-1,2-Bis ( (2S,5S)-2,5-diethylphospholano)benzene (cyclooctadiene)rhodium (I) trifluoromethanesulfonate

    • Packaged under Argon

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit