Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis[(2S,5S)-2,5-diisopropyl-1-phospholanyl]benzene, 97+%, Thermo Scientific™

Click to view available options
Quantity:
250mg
1g
147253-69-8
C26H44P2
418.59
MFCD02684550,MFCD02684550
RBVGOQHQBUPSGX-UHFFFAOYNA-N
s,s-i-pr-duphos, rel-1,2-bis 2s,5s-2,5-diisopropylphospholan-1-yl benzene, s,s-ipr-duphos, 1,2-bis 2s,5s-2,5-diisopropylphospholan-1-yl benzene, --1,2-bis 2s,5s-2,5-di-iso-propylphospholano benzene, --1,2-bis 2s,5s-2,5-di-i-propylphospholano benzene, --1,2-bis 2s,5s-2,5-diisopropylphospholano benzene, kanata purity, 2s,5s-1-2-2s,5s-2,5-diisopropylphospholan-1-yl phenyl-2,5-diisopropylphospholane
11520495
(2S,5S)-1-[2-[(2S,5S)-2,5-di(propan-2-yl)phospholan-1-yl]phenyl]-2,5-di(propan-2-yl)phospholane
CC(C)C1CCC(C(C)C)P1C1=CC=CC=C1P1C(CCC1C(C)C)C(C)C
147253-69-8
C26H44P2
418.59
MFCD02684550,MFCD02684550
RBVGOQHQBUPSGX-UHFFFAOYNA-N
s,s-i-pr-duphos, rel-1,2-bis 2s,5s-2,5-diisopropylphospholan-1-yl benzene, s,s-ipr-duphos, 1,2-bis 2s,5s-2,5-diisopropylphospholan-1-yl benzene, --1,2-bis 2s,5s-2,5-di-iso-propylphospholano benzene, --1,2-bis 2s,5s-2,5-di-i-propylphospholano benzene, --1,2-bis 2s,5s-2,5-diisopropylphospholano benzene, kanata purity, 2s,5s-1-2-2s,5s-2,5-diisopropylphospholan-1-yl phenyl-2,5-diisopropylphospholane
11520495
(2S,5S)-1-[2-[(2S,5S)-2,5-di(propan-2-yl)phospholan-1-yl]phenyl]-2,5-di(propan-2-yl)phospholane
CC(C)C1CCC(C(C)C)P1C1=CC=CC=C1P1C(CCC1C(C)C)C(C)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Melting Point 26°C to 43°C
Quantity 250mg
Sensitivity Air Sensitive
Formula Weight 418.58
Optical Rotation −103° (c=1 in Chloroform)
Percent Purity ≥97%
Chemical Name or Material 1,2-Bis[(2S,5S)-2,5-diisopropyl-1-phospholanyl]benzene

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

RUO – Research Use Only