Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis((2S,5S)-2,5-diisopropylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%, ACROS Organics™

Click to view available options
Quantity:
50mg
250mg
1000393-36-1
C34H56P2Rh·BF4
716.48
MFCD07369040
IJKOQQHIAGRJHA-AUXLTOJKSA-N
1,2-bis 2s,5s-2,5-diisopropylphospholano benzene 1,5-cyclooctadiene rhodium i tetrafluoroborate,s,s-i-pr-duphos-rh,--1,2-bis 2s,5s-2,5-di-i-propylphospholano benzene 1,5-cyclooctadiene rhodium i tetrafluoroborate
71310601
(1Z,5Z)-cycloocta-1,5-diene;(2S,5S)-1-[2-[(2S,5S)-2,5-di(propan-2-yl)phospholan-1-yl]phenyl]-2,5-di(propan-2-yl)phospholane;rhodium;tetrafluoroborate
[B-](F)(F)(F)F.CC(C)C1CCC(P1C2=CC=CC=C2P3C(CCC3C(C)C)C(C)C)C(C)C.C1CC=CCCC=C1.[Rh]
1000393-36-1
C34H56P2Rh·BF4
716.48
MFCD07369040
IJKOQQHIAGRJHA-AUXLTOJKSA-N
1,2-bis 2s,5s-2,5-diisopropylphospholano benzene 1,5-cyclooctadiene rhodium i tetrafluoroborate,s,s-i-pr-duphos-rh,--1,2-bis 2s,5s-2,5-di-i-propylphospholano benzene 1,5-cyclooctadiene rhodium i tetrafluoroborate
71310601
(1Z,5Z)-cycloocta-1,5-diene;(2S,5S)-1-[2-[(2S,5S)-2,5-di(propan-2-yl)phospholan-1-yl]phenyl]-2,5-di(propan-2-yl)phospholane;rhodium;tetrafluoroborate
[B-](F)(F)(F)F.CC(C)C1CCC(P1C2=CC=CC=C2P3C(CCC3C(C)C)C(C)C)C(C)C.C1CC=CCCC=C1.[Rh]
Infrared Spectrum Authentic
Quantity 50mg
NMR (1H-NMR) Consistent with structure, (31P-NMR) Consistent with structure, NMR (19F-NMR), Consistent with structure
Color Brown to Yellow
Elemental Analysis 54.7 to 59.4% C, 7.6 to 8.2% H
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 716.48
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 1,2-Bis((2S,5S)-2,5-Diisopropylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate
Show More Show Less