Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-1,2-Bis[(2S,5S)-diethylphospholano)benzene(cyclooctadiene]rhodium(I) triflate, 98%, ACROS Organics™

Click to view available options
Quantity:
100mg
Packaging:
Glass bottle
142184-30-3
C31H48F3O3P2RhS-
722.63
MFCD00269861
XGPXBCKGQLCHDW-ZCTOJWETSA-M
1,2-bis 2s,5s-2,5-diethylphospholano benzene 1,5-cyclooctadiene rhodium i trifluoromethanesulfonate,+-1,2-bis 2s,5s-2,5-diethylphospholano benzene 1,5-cyclooctadiene rhodium i trifluoromethanesulfonate,2s,5s-1-2-2s,5s-2,5-diethylphospholan-1-yl phenyl-2,5-diethylphospholane 1,5-cyclooctadiene, z,z-rhodium triflate,1,2-bis 2s,5s-2,5-diethyl-phospholano benzene 1,5-cyclooctadiene rhodium i trifluoromethanesulfonate
11146978
(1Z,5Z)-cycloocta-1,5-diene;(2S,5S)-1-[2-[(2S,5S)-2,5-diethylphospholan-1-yl]phenyl]-2,5-diethylphospholane;rhodium;trifluoromethanesulfonate
CCC1CCC(P1C2=CC=CC=C2P3C(CCC3CC)CC)CC.C1CC=CCCC=C1.C(F)(F)(F)S(=O)(=O)[O-].[Rh]
142184-30-3
C31H48F3O3P2RhS-
722.63
MFCD00269861
XGPXBCKGQLCHDW-ZCTOJWETSA-M
1,2-bis 2s,5s-2,5-diethylphospholano benzene 1,5-cyclooctadiene rhodium i trifluoromethanesulfonate,+-1,2-bis 2s,5s-2,5-diethylphospholano benzene 1,5-cyclooctadiene rhodium i trifluoromethanesulfonate,2s,5s-1-2-2s,5s-2,5-diethylphospholan-1-yl phenyl-2,5-diethylphospholane 1,5-cyclooctadiene, z,z-rhodium triflate,1,2-bis 2s,5s-2,5-diethyl-phospholano benzene 1,5-cyclooctadiene rhodium i trifluoromethanesulfonate
11146978
(1Z,5Z)-cycloocta-1,5-diene;(2S,5S)-1-[2-[(2S,5S)-2,5-diethylphospholan-1-yl]phenyl]-2,5-diethylphospholane;rhodium;trifluoromethanesulfonate
CCC1CCC(P1C2=CC=CC=C2P3C(CCC3CC)CC)CC.C1CC=CCCC=C1.C(F)(F)(F)S(=O)(=O)[O-].[Rh]
Infrared Spectrum Authentic
Color Orange to Orange-Red
Quantity 100mg
Elemental Analysis 97.5% min.
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 722.63
Physical Form Crystals
Chemical Name or Material (+)-1, 2-Bis[(2S, 5S)-diethylphospholano)benzene(cyclooctadiene]rhodium(I) triflate
Show More Show Less