Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis(dicyclohexylphosphino)ethane, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
23743-26-2
C26H48P2
422.61
MFCD00015521
BOUYBUIVMHNXQB-UHFFFAOYSA-N
1,2-bis dicyclohexylphosphino ethane,ethylenebis dicyclohexylphosphine,phosphine, 1,2-ethanediylbis dicyclohexyl,1,2-ethanediylbis dicyclohexyl phosphine,dicyclohexyl 2-dicyclohexylphosphanylethyl phosphane,bis 1,2-dicyclohexylphosphino ethane,pubchem6558,acmc-1capw,ethane-1,2-diylbis dicyclohexylphosphane,phosphine, 1,2-ethanediylbis*dicyclohexyl
534202
dicyclohexyl(2-dicyclohexylphosphanylethyl)phosphane
C1CCC(CC1)P(CCP(C2CCCCC2)C3CCCCC3)C4CCCCC4
23743-26-2
C26H48P2
422.61
MFCD00015521
BOUYBUIVMHNXQB-UHFFFAOYSA-N
1,2-bis dicyclohexylphosphino ethane,ethylenebis dicyclohexylphosphine,phosphine, 1,2-ethanediylbis dicyclohexyl,1,2-ethanediylbis dicyclohexyl phosphine,dicyclohexyl 2-dicyclohexylphosphanylethyl phosphane,bis 1,2-dicyclohexylphosphino ethane,pubchem6558,acmc-1capw,ethane-1,2-diylbis dicyclohexylphosphane,phosphine, 1,2-ethanediylbis*dicyclohexyl
534202
dicyclohexyl(2-dicyclohexylphosphanylethyl)phosphane
C1CCC(CC1)P(CCP(C2CCCCC2)C3CCCCC3)C4CCCCC4
Infrared Spectrum Authentic
Quantity 1g
Packaging Glass bottle
Color White
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min
Solubility Information Solubility in water: insoluble in water
Formula Weight 422.61
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 1, 2-Bis(dicyclohexylphosphino)ethane
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA