Alfa Aesar™ 1,2-Bis(diethylphosphino)ethane, 98%

Manufacturer: Alfa Aesar™ 3927203

Quantity 1g

 View more versions of this product

Catalog No. AA3927203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6411-21-8
    Linear Formula (CH3CH2)2PCH2CH2P(CH2CH3)2
    Formula Weight 206.25
    MDL Number MFCD00015175
    UN Number UN2845
    Chemical Name or Material 1,2-Bis(diethylphosphino)ethane

    Specifications

    Formula Weight 206.25
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P361-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Store contents under inert gas.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00015175
    Boiling Point 124° to 126°C (10mmHg)
    Physical Form Liquid
    UN Number UN2845
    Density 0.884g/mL
    Color Colorless
    Quantity 1g
    Odor Pungent
    Sensitivity Air Sensitive
    Linear Formula (CH3CH2)2PCH2CH2P(CH2CH3)2
    CAS 6411-21-8
    Health Hazard 2 GHS H Statement
    H250-H252-H301-H311-H331
    Catches fire spontaneously if exposed to air.
    Self-heating in large quantities
    may catch fire.
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.
    Health Hazard 1 Danger
    Storage Note 1
    • Packaged under argon
    Chemical Name or Material 1,2-Bis(diethylphosphino)ethane
    Assay 98%

    GHS H Statement
    H250-H252-H301-H311-H331
    Catches fire spontaneously if exposed to air.
    Self-heating in large quantities
    may catch fire.
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.

    GHS P Statement
    P210-P301+P310-P361-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Store contents under inert gas.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit