Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis(dipentafluorophenylphosphino)ethane, 95%, Acros Organics™

Click to view available options
Quantity:
1g
5g
76858-94-1
C26H4F20P2
758.26
IGLFIYOFKVGEBP-UHFFFAOYSA-N
1,2-bis dipentafluorophenylphosphino ethane,1,2-bis bis pentafluorophenyl phosphino ethane,1,2-bis bis perfluorophenyl phosphino ethane,2-bis 2,3,4,5,6-pentafluorophenyl phosphanylethyl-bis 2,3,4,5,6-pentafluorophenyl phosphane,ethane-1,2-diylbis bis pentafluorophenyl phosphane,dfppe,pubchem6546,acmc-209p6f,ethylenebis bis pentafluorophenyl phosphine,bis dipentafluorophenylphosphine ethane
2734566
2-bis(2,3,4,5,6-pentafluorophenyl)phosphanylethyl-bis(2,3,4,5,6-pentafluorophenyl)phosphane
C(CP(C1=C(C(=C(C(=C1F)F)F)F)F)C2=C(C(=C(C(=C2F)F)F)F)F)P(C3=C(C(=C(C(=C3F)F)F)F)F)C4=C(C(=C(C(=C4F)F)F)F)F
76858-94-1
C26H4F20P2
758.26
IGLFIYOFKVGEBP-UHFFFAOYSA-N
1,2-bis dipentafluorophenylphosphino ethane,1,2-bis bis pentafluorophenyl phosphino ethane,1,2-bis bis perfluorophenyl phosphino ethane,2-bis 2,3,4,5,6-pentafluorophenyl phosphanylethyl-bis 2,3,4,5,6-pentafluorophenyl phosphane,ethane-1,2-diylbis bis pentafluorophenyl phosphane,dfppe,pubchem6546,acmc-209p6f,ethylenebis bis pentafluorophenyl phosphine,bis dipentafluorophenylphosphine ethane
2734566
2-bis(2,3,4,5,6-pentafluorophenyl)phosphanylethyl-bis(2,3,4,5,6-pentafluorophenyl)phosphane
C(CP(C1=C(C(=C(C(=C1F)F)F)F)F)C2=C(C(=C(C(=C2F)F)F)F)F)P(C3=C(C(=C(C(=C3F)F)F)F)F)C4=C(C(=C(C(=C4F)F)F)F)F
Color White to Beige
Quantity 5g
Melting Point 190.0°C to 195.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Solubility Information Solubility in water: insoluble in water
Formula Weight 758.26
Physical Form Crystalline Powder
Percent Purity 95%
Chemical Name or Material 1, 2-Bis(dipentafluorophenylphosphino)ethane

GHS Signal Word: Warning