Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis(diphenylphosphino)ethane nickel(II) chloride, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
14647-23-5
C26H24Cl2NiP2
528.02
MFCD00013313
XXECWTBMGGXMKP-UHFFFAOYSA-N
1,2-bis diphenylphosphino ethane nickel ii chloride
131664338
C1=CC=C(C=C1)P(CCP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4.Cl.Cl.[Ni+2]
14647-23-5
C26H24Cl2NiP2
528.02
MFCD00013313
XXECWTBMGGXMKP-UHFFFAOYSA-N
1,2-bis diphenylphosphino ethane nickel ii chloride
131664338
C1=CC=C(C=C1)P(CCP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4.Cl.Cl.[Ni+2]
Infrared Spectrum Authentic
Quantity 1g
Color Orange to Red
Packaging Glass bottle
Elemental Analysis 98.5% min.
Melting Point 263.0°C to 265.0°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Solubility Information Solubility in water: insoluble in water
Formula Weight 528.02
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 1, 2-Bis(diphenylphosphino)ethane nickel(II) chloride
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Suspected of causing cancer.

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning