Alfa Aesar™ 1,2-Bis(diphenylphosphinomethyl)benzene, 95%

Manufacturer: Alfa Aesar™ B2508806

Quantity 5g

 View more versions of this product

Catalog No. AAB2508806

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 62144-65-4
    Molecular Formula C32H28P2
    Formula Weight 474.53
    MDL Number MFCD00015429
    Synonym alpha,alpha'-Bis(diphenylphosphino)-o-xylene
    Chemical Name or Material 1,2-Bis(diphenylphosphinomethyl)benzene

    Specifications

    Formula Weight 474.53
    MDL Number MFCD00015429
    Synonym alpha,alpha'-Bis(diphenylphosphino)-o-xylene
    Melting Point 123° to 125°C
    Health Hazard 1 Warning
    Quantity 5g
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C32H28P2
    CAS 62144-65-4
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material 1,2-Bis(diphenylphosphinomethyl)benzene
    Assay 95%

    Description

    Applications
    1,2-Bis(diphenylphosphinomethyl)benzene is used as a bidenate ligand in coordination chemistry and form complexes with transition metal. It reacts with silver salts viz. silver chloride, silver iodide and silver nitrate.

    Notes
    Incompatible with strong oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit