Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis(trimethylsilyloxy)ethane, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
7381-30-8
C8H22O2Si2
206.43
MFCD00009640
JGWFUSVYECJQDT-UHFFFAOYSA-N
1,2-bis trimethylsiloxy ethane,1,2-bis trimethylsilyloxy ethane,2,2,7,7-tetramethyl-3,6-dioxa-2,7-disilaoctane,ethylene glycol bis trimethylsilyl ether,ethylenedioxybis trimethylsilane,3,6-dioxa-2,7-disilaoctane, 2,2,7,7-tetramethyl,1,2-bis-trimethylsilyloxy ethane,1,2-bis-trimethylsilanyloxy-ethane,ethylene glycol, di-tms,acmc-1bh50
81858
trimethyl(2-trimethylsilyloxyethoxy)silane
C[Si](C)(C)OCCO[Si](C)(C)C
7381-30-8
C8H22O2Si2
206.43
MFCD00009640
JGWFUSVYECJQDT-UHFFFAOYSA-N
1,2-bis trimethylsiloxy ethane,1,2-bis trimethylsilyloxy ethane,2,2,7,7-tetramethyl-3,6-dioxa-2,7-disilaoctane,ethylene glycol bis trimethylsilyl ether,ethylenedioxybis trimethylsilane,3,6-dioxa-2,7-disilaoctane, 2,2,7,7-tetramethyl,1,2-bis-trimethylsilyloxy ethane,1,2-bis-trimethylsilanyloxy-ethane,ethylene glycol, di-tms,acmc-1bh50
81858
trimethyl(2-trimethylsilyloxyethoxy)silane
C[Si](C)(C)OCCO[Si](C)(C)C
Boiling Point 165°C to 166°C
Flash Point 46°C
Specific Gravity 0.84
Refractive Index 1.4020 to 1.404
Density 0.8400g/mL
Color Colorless to Yellow
Quantity 50g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (CH3)3SiOCH2CH2OSi(CH3)3
Formula Weight 206.43
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 1, 2-Bis(trimethylsilyloxy)ethane
Show More Show Less

GHS H Statement
Flammable liquid and vapour.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.

GHS Signal Word: Warning

EINECSNumber : 230-950-3