Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Bromoethoxy)-2-bromobenzene, 95%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
18800-28-7
C8H8Br2O
279.96
MFCD00634301
LOVVQTBEUIBASP-UHFFFAOYSA-N
1-bromo-2-2-bromoethoxy benzene,1-2-bromoethoxy-2-bromobenzene,o-bromo-2-bromoethoxybenzene,benzene, 1-bromo-2-2-bromoethoxy
2734928
1-bromo-2-(2-bromoethoxy)benzene
C1=CC=C(C(=C1)OCCBr)Br
18800-28-7
C8H8Br2O
279.96
MFCD00634301
LOVVQTBEUIBASP-UHFFFAOYSA-N
1-bromo-2-2-bromoethoxy benzene,1-2-bromoethoxy-2-bromobenzene,o-bromo-2-bromoethoxybenzene,benzene, 1-bromo-2-2-bromoethoxy
2734928
1-bromo-2-(2-bromoethoxy)benzene
C1=CC=C(C(=C1)OCCBr)Br
Infrared Spectrum Authentic
Quantity 5g
Melting Point 35.0°C to 36.0°C
Color Colorless to Yellow
Boiling Point 160.0°C to 162.0°C (16.0mmHg)
Packaging Glass bottle
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Linear Formula BrC6H4O(CH2)2Br
Formula Weight 279.96
Physical Form Liquid After Melting
Percent Purity 95%
Chemical Name or Material 1-(2-Bromoethoxy)-2-bromobenzene
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Recommended Storage : Product may darken during storage