Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Bromoethoxy)-3-nitrobenzene, 97%, Acros Organics

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
13831-59-9
C8H8BrNO3
246.06
MFCD07783648
QBUSKXLDUNPEMZ-UHFFFAOYSA-N
1-2-bromoethoxy-3-nitrobenzene,benzene,1-2-bromoethoxy-3-nitro,2-bromoethyl 3-nitrophenyl ether,acmc-209chl,2-bromo-3'-nitrophenetole,2-3-nitrophenoxy ethyl bromide,3-2-bromoethoxy-1-nitrobenzene
269626
1-(2-bromoethoxy)-3-nitrobenzene
C1=CC(=CC(=C1)OCCBr)[N+](=O)[O-]
13831-59-9
C8H8BrNO3
246.06
MFCD07783648
QBUSKXLDUNPEMZ-UHFFFAOYSA-N
1-2-bromoethoxy-3-nitrobenzene,benzene,1-2-bromoethoxy-3-nitro,2-bromoethyl 3-nitrophenyl ether,acmc-209chl,2-bromo-3'-nitrophenetole,2-3-nitrophenoxy ethyl bromide,3-2-bromoethoxy-1-nitrobenzene
269626
1-(2-bromoethoxy)-3-nitrobenzene
C1=CC(=CC(=C1)OCCBr)[N+](=O)[O-]
Quantity 25g
Boiling Point 140°C to 145°C (2.0mmHg)
Refractive Index 1.5830 to 1.585
Melting Point 39°C
Packaging Glass bottle
Infrared Spectrum Authentic
Color Brown or Green-Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula NO2C6H4OCH2CH2Br
Formula Weight 246.06
Physical Form Low Melting Solid
Percent Purity ≥96.0%
Chemical Name or Material 1-(2-Bromoethoxy)-3-nitrobenzene
Show More Show Less

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.