Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Bromoethoxy-4-bromobenzene) 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
18800-30-1
C8H8Br2O
279.959
MFCD00044201
SRGBSEMMGLSUMI-UHFFFAOYSA-N
1-bromo-4-2-bromoethoxy benzene,2-bromoethyl p-bromophenyl ether,4-bromo-1-2-bromoethoxy benzene,2-bromoethyl 4-bromophenyl ether,benzene,1-bromo-4-2-bromoethoxy,benzene, 1-bromo-4-2-bromoethoxy,4-bromophenoxyethyl bromide,phenetole, p,.beta.-dibromo,2-4-bromophenoxy ethylbromide,2-p-bromophenoxy ethyl bromide
87797
1-bromo-4-(2-bromoethoxy)benzene
C1=CC(=CC=C1OCCBr)Br
18800-30-1
C8H8Br2O
279.959
MFCD00044201
SRGBSEMMGLSUMI-UHFFFAOYSA-N
1-bromo-4-2-bromoethoxy benzene,2-bromoethyl p-bromophenyl ether,4-bromo-1-2-bromoethoxy benzene,2-bromoethyl 4-bromophenyl ether,benzene,1-bromo-4-2-bromoethoxy,benzene, 1-bromo-4-2-bromoethoxy,4-bromophenoxyethyl bromide,phenetole, p,.beta.-dibromo,2-4-bromophenoxy ethylbromide,2-p-bromophenoxy ethyl bromide
87797
1-bromo-4-(2-bromoethoxy)benzene
C1=CC(=CC=C1OCCBr)Br
Color White to Beige
Boiling Point 163.0°C to 165.0°C (16.0mmHg)
Packaging Glass bottle
Infrared Spectrum Authentic
Flash Point 180°C
Quantity 25g
Melting Point 56.0°C to 57.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula BrC6H4OCH2CH2Br
Beilstein 06, I, 105
Formula Weight 279.96
Physical Form Crystalline Powder or Crystals
Percent Purity 98%
Chemical Name or Material 1-(2-Bromoethoxy-4-bromobenzene)
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.