Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-(2-Bromoethyl)-3-(trifluoromethyl)benzene, 95%

Click to view available options
Quantity:
250mg
1g
5g
1997-80-4
C9H8BrF3
253.062
MFCD03092900
AZLYAPKPZBXCGX-UHFFFAOYSA-N
1-2-bromoethyl-3-trifluoromethyl benzene,3-trifluoromethyl phenethyl bromide,3-trifluoromethyl phenylethyl bromide,benzene, 1-2-bromoethyl-3-trifluoromethyl,1-bromo-2-3-trifluoromethyl phenyl ethane,acmc-20anht,3-trifluoromethylphenethyl bromide,toluene, m-2-bromoethyl-.alpha.,.alpha.,.alpha.-trifluoro,3-trifluoromethyl phenylethylbromide,3-trifluoromethyl phenethyl bromide
594535
1-(2-bromoethyl)-3-(trifluoromethyl)benzene
C1=CC(=CC(=C1)C(F)(F)F)CCBr
1997-80-4
C9H8BrF3
253.062
MFCD03092900
AZLYAPKPZBXCGX-UHFFFAOYSA-N
1-2-bromoethyl-3-trifluoromethyl benzene,3-trifluoromethyl phenethyl bromide,3-trifluoromethyl phenylethyl bromide,benzene, 1-2-bromoethyl-3-trifluoromethyl,1-bromo-2-3-trifluoromethyl phenyl ethane,acmc-20anht,3-trifluoromethylphenethyl bromide,toluene, m-2-bromoethyl-.alpha.,.alpha.,.alpha.-trifluoro,3-trifluoromethyl phenylethylbromide,3-trifluoromethyl phenethyl bromide
594535
1-(2-bromoethyl)-3-(trifluoromethyl)benzene
C1=CC(=CC(=C1)C(F)(F)F)CCBr
Density 1.495g/mL
Quantity 250mg
Refractive Index 1.486
Color Colorless
Boiling Point 92°C to 94°C (12mmHg)
UN Number UN3082
Beilstein 1950052
Formula Weight 253.06
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 1-(2-Bromoethyl)-3-(trifluoromethyl)benzene
Show More Show Less

GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P301+P312-P337+P313-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.