Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-(2-Chloro-4-nitrophenyl)-4-methylpiperazine, 97%

Click to view available options
Quantity:
250mg
1g
5g
16154-62-4
C11H14ClN3O2
255.702
MFCD00030787
UFFIYCVVTWWGGH-UHFFFAOYSA-N
1-2-chloro-4-nitrophenyl-4-methylpiperazine,1-2-chloro-4-nitro-phenyl-4-methyl-piperazine,1-methyl-4-2-chloro-4-nitrophenyl piperazine,1-2-chloranyl-4-nitro-phenyl-4-methyl-piperazine
2837294
CN1CCN(CC1)C2=C(C=C(C=C2)[N+](=O)[O-])Cl
16154-62-4
C11H14ClN3O2
255.702
MFCD00030787
UFFIYCVVTWWGGH-UHFFFAOYSA-N
1-2-chloro-4-nitrophenyl-4-methylpiperazine,1-2-chloro-4-nitro-phenyl-4-methyl-piperazine,1-methyl-4-2-chloro-4-nitrophenyl piperazine,1-2-chloranyl-4-nitro-phenyl-4-methyl-piperazine
2837294
CN1CCN(CC1)C2=C(C=C(C=C2)[N+](=O)[O-])Cl
Quantity 1g
Formula Weight 255.7
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 1-(2-Chloro-4-nitrophenyl)-4-methylpiperazine

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures