Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-(2-Chloro-4-nitrophenyl)piperidine, 97%

Click to view available options
Quantity:
250mg
1g
5g
55403-25-3
C11H13ClN2O2
240.687
AIZKHORWEJERNY-UHFFFAOYSA-N
1-2-chloro-4-nitrophenyl piperidine,1-2-chloro-4-nitro-phenyl-piperidine,2-chloro-4-nitrophenyl piperidine,maybridge1_000868,piperidine, 1-2-chloro-4-nitrophenyl
2757742
1-(2-chloro-4-nitrophenyl)piperidine
C1CCN(CC1)C2=C(C=C(C=C2)[N+](=O)[O-])Cl
55403-25-3
C11H13ClN2O2
240.687
AIZKHORWEJERNY-UHFFFAOYSA-N
1-2-chloro-4-nitrophenyl piperidine,1-2-chloro-4-nitro-phenyl-piperidine,2-chloro-4-nitrophenyl piperidine,maybridge1_000868,piperidine, 1-2-chloro-4-nitrophenyl
2757742
1-(2-chloro-4-nitrophenyl)piperidine
C1CCN(CC1)C2=C(C=C(C=C2)[N+](=O)[O-])Cl
Quantity 1g
Formula Weight 240.69
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 1-(2-Chloro-4-nitrophenyl)piperidine

TSCA : No

Recommended Storage : Ambient temperatures