Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 1-(2-Chloro-4-nitrophenyl)piperidine, 97%
Click to view available options
Quantity:
250mg
1g
5g
Specifications
Quantity | 250mg |
Formula Weight | 240.69 |
Physical Form | Solid |
Percent Purity | 97% |
Chemical Name or Material | 1-(2-Chloro-4-nitrophenyl)piperidine |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures