Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-(2-Chloro-6-nitrophenyl)azepane, 97%

Click to view available options
Quantity:
250mg
1g
5g
1-(2-Chloro-6-nitrophenyl)hexamethyleneimine
1-(2-Chloro-6-nitrophenyl)hexamethyleneimine
Quantity 5g
UN Number UN3259
Formula Weight 254.71
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 1-(2-Chloro-6-nitrophenyl)azepane

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures