Alfa Aesar™ 1-(2-Chloroethyl)piperidine hydrochloride, 98%

Manufacturer: Alfa Aesar™ B2298536

Quantity 500g

 View more versions of this product

Catalog No. AAB2298536

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2008-75-5
    Molecular Formula C7H14ClN·HCl
    Formula Weight 184.11
    MDL Number MFCD00012837
    UN Number UN2928
    Beilstein 3910268
    Synonym 2-(1-Piperidino)ethyl chloride hydrochloride
    Chemical Name or Material 1-(2-Chloroethyl)piperidine hydrochloride

    Specifications

    Formula Weight 184.11
    MDL Number MFCD00012837
    CAS 2008-75-5
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, CORROSIVE, ORGANIC, N.O.S.
    Melting Point 229° to 235°C
    Quantity 500g
    Molecular Formula C7H14ClN·HCl
    UN Number UN2928
    Beilstein 3910268
    Synonym 2-(1-Piperidino)ethyl chloride hydrochloride
    Health Hazard 2 GHS H Statement
    H300-H314-H318-H341
    Fatal if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Suspected of causing genetic defects.
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number TM6482500
    Chemical Name or Material 1-(2-Chloroethyl)piperidine hydrochloride
    Assay 98%

    GHS H Statement
    H300-H314-H318-H341
    Fatal if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Suspected of causing genetic defects.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit