Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-(2-Chloroethyl)pyrrolidine hydrochloride, 98%

Click to view available options
Quantity:
25g
100g
7250-67-1
C6H13Cl2N
170.077
MFCD00012718
FSNGFFWICFYWQC-UHFFFAOYSA-N
1-2-chloroethyl pyrrolidine hydrochloride,n-2-chloroethyl pyrrolidine hydrochloride,2-chloroethylpyrrolidine hydrochloride,2-pyrrolidinoethyl chloride hydrochloride,pyrrolidine, 1-2-chloroethyl-, hydrochloride,unii-yyq1h343r5,pyrrolidinoethyl chloride, hydrochloride,1-2-chloroethyl pyrrolidine.hcl,1-2-chloroethyl pyrrolidine hcl,2-chloroethylpyrrolidinehydrochloride
81668
1-(2-chloroethyl)pyrrolidine;hydrochloride
C1CCN(C1)CCCl.Cl
7250-67-1
C6H13Cl2N
170.077
MFCD00012718
FSNGFFWICFYWQC-UHFFFAOYSA-N
1-2-chloroethyl pyrrolidine hydrochloride,n-2-chloroethyl pyrrolidine hydrochloride,2-chloroethylpyrrolidine hydrochloride,2-pyrrolidinoethyl chloride hydrochloride,pyrrolidine, 1-2-chloroethyl-, hydrochloride,unii-yyq1h343r5,pyrrolidinoethyl chloride, hydrochloride,1-2-chloroethyl pyrrolidine.hcl,1-2-chloroethyl pyrrolidine hcl,2-chloroethylpyrrolidinehydrochloride
81668
1-(2-chloroethyl)pyrrolidine;hydrochloride
C1CCN(C1)CCCl.Cl
Quantity 100g
Melting Point 168°C to 173°C
Beilstein 3677061
Formula Weight 170.08
Percent Purity 98%
Chemical Name or Material 1-(2-Chloroethyl)pyrrolidine hydrochloride

GHS H Statement
H301-H332-H315-H319-H335
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : UY0703150