Alfa Aesar™ 1-(2-Chlorophenyl)ethanol, 96%

Manufacturer: Alfa Aesar™ B2480022

Quantity 100g

 View more versions of this product

Catalog No. AAB2480022

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13524-04-4
    Molecular Formula C8H9ClO
    Formula Weight 156.61
    MDL Number MFCD00041037
    Synonym 2-Chlorophenyl methyl carbinol; 2-Chloro-alpha-methylbenzyl alcohol
    Chemical Name or Material 1-(2-Chlorophenyl)ethanol

    Specifications

    Formula Weight 156.61
    TSCA TSCA
    MDL Number MFCD00041037
    Molecular Formula C8H9ClO
    CAS 13524-04-4
    Synonym 2-Chlorophenyl methyl carbinol; 2-Chloro-alpha-methylbenzyl alcohol
    Boiling Point 121° to 123°C (14mmHg)
    Quantity 100g
    Chemical Name or Material 1-(2-Chlorophenyl)ethanol
    Assay 96%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit