Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1,2-Cyclohexylenedinitrilo)tetraacetic Acid, Reagent, 98%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
12 kg
Packaging:
Amber Glass Bottle
Poly Pail
125572-95-4
C14H20N2O8
344.32
FCKYPQBAHLOOJQ-NXEZZACHSA-L
2-{[(1R,2R)-2-[bis(carboxylatomethyl)azaniumyl]cyclohexyl](carboxylatomethyl)azaniumyl}acetate
[O-]C(=O)C[NH+](CC([O-])=O)[C@@H]1CCCC[C@H]1[NH+](CC([O-])=O)CC([O-])=O
125572-95-4
C14H20N2O8
344.32
FCKYPQBAHLOOJQ-NXEZZACHSA-L
2-{[(1R,2R)-2-[bis(carboxylatomethyl)azaniumyl]cyclohexyl](carboxylatomethyl)azaniumyl}acetate
[O-]C(=O)C[NH+](CC([O-])=O)[C@@H]1CCCC[C@H]1[NH+](CC([O-])=O)CC([O-])=O

C1466, 125572-95-4, C14H22N2O8•H2O

Spectrum™ (1,2-Cyclohexylenedinitrilo)tetraacetic Acid, Reagent is an alkyl-substituted amino acid that functions as a chelating agent. This ingredient′s source is synthetic. It appears as a solid and is partially soluble in cold water. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.
CAS Max % 1
Percent Purity 98%
Packaging Amber Glass Bottle
Quantity 500 g