Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Decanediol, 98%, ACROS Organics™

Click to view available options
Quantity:
50g
250g
Packaging:
Glass bottle
1119-86-4
C10H22O2
174.284
MFCD00010739
YSRSBDQINUMTIF-UHFFFAOYSA-N
1,2-decanediol,decylene glycol,decane 1,2-diol,1,2-dihydroxydecane,acmc-20apfz,acmc-20apjb,pubchem12680,acmc-1bny3
79141
decane-1,2-diol
CCCCCCCCC(CO)O
1119-86-4
C10H22O2
174.284
MFCD00010739
YSRSBDQINUMTIF-UHFFFAOYSA-N
1,2-decanediol,decylene glycol,decane 1,2-diol,1,2-dihydroxydecane,acmc-20apfz,acmc-20apjb,pubchem12680,acmc-1bny3
79141
decane-1,2-diol
CCCCCCCCC(CO)O
Melting Point 47°C to 51°C
Color White
Quantity 50g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3(CH2)7CH(OH)CH2OH
Formula Weight 174.28
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 1, 2-Decanediol
Show More Show Less