Alfa Aesar™ 1,2-Di(3-indenyl)ethane, 98%

Manufacturer: Alfa Aesar™ H2719503

Quantity 1g

 View more versions of this product

Catalog No. AAH2719503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 18657-57-3
    Molecular Formula C20H18
    Formula Weight 258.36
    MDL Number MFCD00191814
    Chemical Name or Material 1,2-Di(3-indenyl)ethane

    Specifications

    Formula Weight 258.36
    TSCA TSCA
    Melting Point 121° to 125°C
    Molecular Formula C20H18
    Quantity 1g
    MDL Number MFCD00191814
    CAS 18657-57-3
    Chemical Name or Material 1,2-Di(3-indenyl)ethane
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit