Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Diamino-4,5-difluorobenzene, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
25g
76179-40-3
C6H6F2N2
144.12
PPWRHKISAQTCCG-UHFFFAOYSA-N
1,2-diamino-4,5-difluorobenzene,4,5-difluoro-1,2-phenylenediamine,4,5-difluoro-o-phenylenediamine,2-amino-4,5-difluorophenylamine,4,5-difluoro-benzene-1,2-diamine,4,5-difluoro-1,2-benzenediamine,1,2-benzenediamine,4,5-difluoro,1,2-benzenediamine, 4,5-difluoro,pubchem4354,2-amino-4,5-difluoroaniline
2736755
4,5-difluorobenzene-1,2-diamine
C1=C(C(=CC(=C1F)F)N)N
76179-40-3
C6H6F2N2
144.12
PPWRHKISAQTCCG-UHFFFAOYSA-N
1,2-diamino-4,5-difluorobenzene,4,5-difluoro-1,2-phenylenediamine,4,5-difluoro-o-phenylenediamine,2-amino-4,5-difluorophenylamine,4,5-difluoro-benzene-1,2-diamine,4,5-difluoro-1,2-benzenediamine,1,2-benzenediamine,4,5-difluoro,1,2-benzenediamine, 4,5-difluoro,pubchem4354,2-amino-4,5-difluoroaniline
2736755
4,5-difluorobenzene-1,2-diamine
C1=C(C(=CC(=C1F)F)N)N
Melting Point 130°C to 133°C
Quantity 1g
Packaging Glass bottle
Assay Percent Range 96% min. (GC)
Linear Formula (F)C6H2(NH2)2
Formula Weight 144.12
Percent Purity 97%
Chemical Name or Material 1, 2-Diamino-4, 5-difluorobenzene

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning