Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Diaminocyclohexane, mixture of isomers, 99%, Thermo Scientific Chemicals

Catalog Number AAB246570B
Click to view available options
Quantity:
250 g
1000 g
694-83-7
C6H14N2
114.192
MFCD00001491
SSJXIUAHEKJCMH-UHFFFAOYSA-N
1,2-cyclohexanediamine, 1,2-diaminocyclohexane, 1,2-cylohexanediamine, cyclohex-1,2-ylenediamine, 1r,2r---1,2-diamino cyclohexane, dsstox_cid_7301, dsstox_rid_78396, dsstox_gsid_27301, +/--trans-1,2-diaminocyclohexane, cis-1,2-cyclohexandiamine
4610
cyclohexane-1,2-diamine
C1CCC(C(C1)N)N
694-83-7
C6H14N2
114.192
MFCD00001491
SSJXIUAHEKJCMH-UHFFFAOYSA-N
1,2-cyclohexanediamine, 1,2-diaminocyclohexane, 1,2-cylohexanediamine, cyclohex-1,2-ylenediamine, 1r,2r---1,2-diamino cyclohexane, dsstox_cid_7301, dsstox_rid_78396, dsstox_gsid_27301, +/--trans-1,2-diaminocyclohexane, cis-1,2-cyclohexandiamine
4610
cyclohexane-1,2-diamine
C1CCC(C(C1)N)N

1,2-Diaminocyclohexane was used in the synthesis of chiral ruthenium(IV)-oxo complexes & bicyclic pyrazine derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Diaminocyclohexane was used in the synthesis of chiral ruthenium(IV)-oxo complexes & bicyclic pyrazine derivatives.

Solubility
Fully miscible in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents, air, carbon dioxide.
TRUSTED_SUSTAINABILITY
Density 0.94
pH 12
Boiling Point 191°C
Flash Point 75°C (167°F)
Odor Amine-like
Refractive Index 1.487
Quantity 1000 g
UN Number UN2735
Beilstein 506142
Sensitivity Air Sensitive
Solubility Information Fully miscible in water.
Formula Weight 114.19
Percent Purity 99%
Chemical Name or Material 1,2-Diaminocyclohexane, mixture of isomers
Show More Show Less
Hazard Category H227-H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318-H317-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 211-776-7
RTECSNumber GU8749500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit