Alfa Aesar™ 1,2-Diaminopropane, 99%

Manufacturer: Alfa Aesar™ B24489AK

Quantity 250mL

 View more versions of this product

Catalog No. AAB24489AK

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 78-90-0
    Molecular Formula C3H10N2
    Formula Weight 74.13
    MDL Number MFCD00008089
    UN Number UN2258
    Beilstein 605274
    Merck Index 14,7852
    Synonym 1,2-Propanediamine; Propylenediamine
    Chemical Name or Material 1,2-Diaminopropane

    Specifications

    Formula Weight 74.13
    Odor Characteristic
    Molecular Formula C3H10N2
    MDL Number MFCD00008089
    Beilstein 605274
    Merck Index 14,7852
    Health Hazard 2 GHS H Statement
    H314-H226-H302-H312
    Causes severe skin burns and eye damage.
    Flammable liquid and vapour.
    Harmful if swallowed.
    Harmful in contact with skin.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: 1,2-PROPYLENEDIAMINE
    Flash Point 34°C (93°F)
    Quantity 250mL
    TSCA TSCA
    CAS 78-90-0
    UN Number UN2258
    Synonym 1,2-Propanediamine; Propylenediamine
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number TX6650000
    Boiling Point 119° to 120°C
    Melting Point ≈−37°C
    Chemical Name or Material 1,2-Diaminopropane
    Assay 99%

    GHS H Statement
    H314-H226-H302-H312
    Causes severe skin burns and eye damage.
    Flammable liquid and vapour.
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit