Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Diaminopropane, 99%, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
2.5L
Packaging:
Glass bottle
78-90-0
C3H10N2
74.13
MFCD00008089
AOHJOMMDDJHIJH-UHFFFAOYSA-N
1,2-diaminopropane,propylenediamine,1,2-propanediamine,propylene diamine,2,3-diaminopropane,1,2-propylenediamine,1,2 diaminopropane,ccris 4863,1-methylethylenediamine,2-aminopropylamine
6567
CHEBI:30630
propane-1,2-diamine
CC(CN)N
78-90-0
C3H10N2
74.13
MFCD00008089
AOHJOMMDDJHIJH-UHFFFAOYSA-N
1,2-diaminopropane,propylenediamine,1,2-propanediamine,propylene diamine,2,3-diaminopropane,1,2-propylenediamine,1,2 diaminopropane,ccris 4863,1-methylethylenediamine,2-aminopropylamine
6567
CHEBI:30630
propane-1,2-diamine
CC(CN)N
Specific Gravity 0.87
Refractive Index 1.4450 to 1.4470
Melting Point -37.0°C
Density 0.8700g/mL
Boiling Point 119.0°C to 120.0°C
Flash Point 33°C
Quantity 250mL
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3CH(NH2)CH2NH2
Beilstein 04,257
Merck Index 15,7965
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in ether, soluble in acetone, benzene, chloroform and, ethanol
Formula Weight 74.13
Percent Purity 99%
Chemical Name or Material 1, 2-Diaminopropane
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger