Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,2-Dibromo-1-iodotrifluoroethane, 97%

Click to view available options
Quantity:
1g
5g
216394-01-3
C2Br2F3I
367.73
MFCD00042521
YUBJHLYGOOZDRN-UHFFFAOYSA-N
1,2-dibromo-1-iodotrifluoroethane,1,2-dibromo-2-iodotrifluoroethane,1,2-dibromo-1,2,2-trifluoro-1-iodoethane,ethane,1,2-dibromo-1,1,2-trifluoro-2-iodo
2736780
1,2-dibromo-1,1,2-trifluoro-2-iodoethane
C(C(F)(Br)I)(F)(F)Br
216394-01-3
C2Br2F3I
367.73
MFCD00042521
YUBJHLYGOOZDRN-UHFFFAOYSA-N
1,2-dibromo-1-iodotrifluoroethane,1,2-dibromo-2-iodotrifluoroethane,1,2-dibromo-1,2,2-trifluoro-1-iodoethane,ethane,1,2-dibromo-1,1,2-trifluoro-2-iodo
2736780
1,2-dibromo-1,1,2-trifluoro-2-iodoethane
C(C(F)(Br)I)(F)(F)Br
Quantity 5g
Boiling Point 63°C to 65°C (32mmHg)
Linear Formula BrCFICF2Br
Formula Weight 367.73
Percent Purity 97%
Chemical Name or Material 1,2-Dibromo-1-iodotrifluoroethane

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger