Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dibromobenzene, 98%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
Packaging:
Glass bottle
583-53-9
C6H4Br2
235.91
MFCD00000057
WQONPSCCEXUXTQ-UHFFFAOYSA-N
o-dibromobenzene,dibromobenzene,benzene, 1,2-dibromo,ortho-dibromobenzene,benzene, o-dibromo,benzene, dibromo,unii-52k9w7u6eh,dibromobenzene mixed isomers,1,2 dibromobenzene,1,6-dibromobenzene
11414
CHEBI:37152
1,2-dibromobenzene
C1=CC=C(C(=C1)Br)Br
583-53-9
C6H4Br2
235.91
MFCD00000057
WQONPSCCEXUXTQ-UHFFFAOYSA-N
o-dibromobenzene,dibromobenzene,benzene, 1,2-dibromo,ortho-dibromobenzene,benzene, o-dibromo,benzene, dibromo,unii-52k9w7u6eh,dibromobenzene mixed isomers,1,2 dibromobenzene,1,6-dibromobenzene
11414
CHEBI:37152
1,2-dibromobenzene
C1=CC=C(C(=C1)Br)Br
Specific Gravity 1.956
Boiling Point 224.0°C
Quantity 100mL
Density 1.9560g/mL
Color Colorless to Yellow or Pink
Packaging Glass bottle
Refractive Index 1.6100 to 1.6120
Infrared Spectrum Authentic
Flash Point 91°C
Melting Point 4.0°C to 6.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula C6H4Br2
Beilstein 05, 210
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol,acetone,ether,benzene,ca,rbon tetrachloride,n-heptane
Formula Weight 235.91
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 1, 2-Dibromobenzene
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN

GHS Signal Word: Warning

EINECSNumber : 209-507-3