Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dibromocyclopentene, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
75415-78-0
C5H6Br2
225.9
PNWFXPGGROADNS-UHFFFAOYSA-N
1,2-dibromocyclopent-1-ene,dibromocyclopentene,pubchem13714,acmc-20ap6k,1,2-bis bromanyl cyclopentene,1,2-dibromocyclopentene
4438528
1,2-dibromocyclopentene
C1CC(=C(C1)Br)Br
75415-78-0
C5H6Br2
225.9
PNWFXPGGROADNS-UHFFFAOYSA-N
1,2-dibromocyclopent-1-ene,dibromocyclopentene,pubchem13714,acmc-20ap6k,1,2-bis bromanyl cyclopentene,1,2-dibromocyclopentene
4438528
1,2-dibromocyclopentene
C1CC(=C(C1)Br)Br
Specific Gravity 1.895
Packaging Glass bottle
Flash Point >110°C
Quantity 1g
Boiling Point 78.0°C (5.0 mmHg)
Density 1.8950g/mL
Assay Percent Range 96% min. (GC)
Formula Weight 225.9
Percent Purity 97%
Chemical Name or Material 1, 2-Dibromocyclopentene

GHS Signal Word: Warning