Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (1,2-Dibromoethyl)benzene, 97%

Click to view available options
Quantity:
25g
100g
500g
93-52-7
C8H8Br2
263.96
MFCD00000138
SHKKTLSDGJRCTR-UHFFFAOYSA-N
1,2-dibromoethyl benzene,dowspray 9,1,2-dibromo-1-phenylethane,1,2-dibromo phenyl ethane,benzene, 1,2-dibromoethyl,1,2-dibromo-2-phenylethane,vic-styrene dibromide,alpha,beta-dibromoethylbenzene,ccris 2090,.alpha.,.beta.-dibromoethylbenzene
7145
1,2-dibromoethylbenzene
C1=CC=C(C=C1)C(CBr)Br
93-52-7
C8H8Br2
263.96
MFCD00000138
SHKKTLSDGJRCTR-UHFFFAOYSA-N
1,2-dibromoethyl benzene,dowspray 9,1,2-dibromo-1-phenylethane,1,2-dibromo phenyl ethane,benzene, 1,2-dibromoethyl,1,2-dibromo-2-phenylethane,vic-styrene dibromide,alpha,beta-dibromoethylbenzene,ccris 2090,.alpha.,.beta.-dibromoethylbenzene
7145
1,2-dibromoethylbenzene
C1=CC=C(C=C1)C(CBr)Br
Boiling Point 139°C to 140°C (15mmHg)
Quantity 25g
Melting Point 70°C to 74°C
UN Number UN3261
Formula Weight 263.96
Percent Purity 97%
Chemical Name or Material (1,2-Dibromoethyl)benzene

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : TSCA