Alfa Aesar™ 1,2-Dibromopropane, 98%

Manufacturer: Alfa Aesar™ B2299436

Quantity 500g

 View more versions of this product

Catalog No. AAB2299436

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 78-75-1
    Molecular Formula C3H6Br2
    Formula Weight 201.9
    MDL Number MFCD00000148
    UN Number UN1993
    Beilstein 1718884
    Merck Index 14,7853
    Synonym Propylene dibromide; Propylene bromide
    Chemical Name or Material 1,2-Dibromopropane

    Specifications

    Formula Weight 201.9
    TSCA TSCA
    MDL Number MFCD00000148
    UN Number UN1993
    Merck Index 14,7853
    Synonym Propylene dibromide; Propylene bromide
    Health Hazard 2 GHS H Statement
    H330-H226-H302-H315-H319
    Fatal if inhaled.
    Flammable liquid and vapour.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point 35°C (95°F)
    Melting Point −55°C
    Molecular Formula C3H6Br2
    CAS 78-75-1
    Beilstein 1718884
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    RTECS Number TX8574000
    Boiling Point 140° to 142°C
    Quantity 500g
    Chemical Name or Material 1,2-Dibromopropane
    Assay 98%

    Description

    Applications
    1,2-Dibromopropane was used to study the effect of reductant concentration, reductant contact time and suspension pH on reductive dechlorination of carbon tetrachloride by soil manipulated with Fe(II) and HS-. It was used as internal standard during the determination of nitrogenous disinfection byproduct trichloronitromethane by gas chromatography mass spectrometry.

    Solubility
    Soluble in water (slightly), ether, and alcohol.

    Notes
    Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

    GHS H Statement
    H330-H226-H302-H315-H319
    Fatal if inhaled.
    Flammable liquid and vapour.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit