Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,2-Dichloro-4-fluoro-5-nitrobenzene, 95%

Click to view available options
Quantity:
5g
25g
100g
2339-78-8
C6H2Cl2FNO2
209.985
MFCD00075330
FXOCDIKCKFOUDE-UHFFFAOYSA-N
4,5-dichloro-2-fluoronitrobenzene,benzene, 1,2-dichloro-4-fluoro-5-nitro,buttpark 89\07-58,4,5-dichloro-2-fluoro-1-nitrobenzene,pubchem4385,intermediates-zcf02626,ksc201k5p,1,2-ichloro-4-luoro-5-itro-benzene,4,5-dichloro-2-fluoronitrobenzene,4,5-dichloro-2-fluoronitrobenzene
533951
1,2-dichloro-4-fluoro-5-nitrobenzene
C1=C(C(=CC(=C1Cl)Cl)F)[N+](=O)[O-]
2339-78-8
C6H2Cl2FNO2
209.985
MFCD00075330
FXOCDIKCKFOUDE-UHFFFAOYSA-N
4,5-dichloro-2-fluoronitrobenzene,benzene, 1,2-dichloro-4-fluoro-5-nitro,buttpark 89\07-58,4,5-dichloro-2-fluoro-1-nitrobenzene,pubchem4385,intermediates-zcf02626,ksc201k5p,1,2-ichloro-4-luoro-5-itro-benzene,4,5-dichloro-2-fluoronitrobenzene,4,5-dichloro-2-fluoronitrobenzene
533951
1,2-dichloro-4-fluoro-5-nitrobenzene
C1=C(C(=CC(=C1Cl)Cl)F)[N+](=O)[O-]
Quantity 5g
Boiling Point 247°C
Color Yellow
Melting Point 17°C
Density 1.596g/mL
Refractive Index 1.5750
Beilstein 3269124
Formula Weight 209.99
Physical Form Low Melting Solid
Percent Purity 95%
Chemical Name or Material 1,2-Dichloro-4-fluoro-5-nitrobenzene
Show More Show Less

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning